1-(3-Bromo-4-chlorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

61849-500MG 61849-1G 61849-5G
77337-568EA 423.33 USD
77337-568 77355-680 77379-726
1-(3-Bromo-4-chlorophenyl)ethanol ≥97%
1-(3-Bromo-4-chlorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD24231593
CAS Number: 1552127-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR