Methyl 2-bromo-4,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

61845-500MG 61845-1G 61845-5G 61845-25G
77331-428EA 206 USD
77331-428 77355-678 77367-590 77389-054
Methyl 2-bromo-4,5-dimethylbenzoate ≥97%
Methyl 2-bromo-4,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD28738481
CAS Number: 1368217-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR