2-Iodo-4,5-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

61844-500MG 61844-1G 61844-5G
77335-870EA 367.71 USD
77335-870 77355-676 77379-724
2-Iodo-4,5-dimethylphenol ≥97%
2-Iodo-4,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉IO
Storage Temperature: Ambient
MDL Number: MFCD07779033
CAS Number: 106362-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR