2-Hydroxy-4,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61838-500MG 61838-1G 61838-5G 61838-25G
77331-426EA 202.91 USD
77331-426 77355-674 77367-588 77389-052
2-Hydroxy-4,5-dimethylbenzoic acid ≥97%
2-Hydroxy-4,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD12196123
CAS Number: 58138-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR