2-(Benzyloxy)-4-ethoxy-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

61834-500MG 61834-1G 61834-5G 61834-25G
77331-420EA 222.48 USD
77331-420 77355-668 77367-582 77389-046
2-(Benzyloxy)-4-ethoxy-1-fluorobenzene ≥97%
2-(Benzyloxy)-4-ethoxy-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₂
MW: 246.2768 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760594
CAS Number: 2764731-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR