1-Bromo-3-chloro-2-iodo-4-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61831-500MG 61831-1G 61831-5G 61831-25G
77331-416EA 188.49 USD
77331-416 77355-664 77367-578 77389-042
1-Bromo-3-chloro-2-iodo-4-propoxybenzene ≥97%
1-Bromo-3-chloro-2-iodo-4-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrClIO
MW: 375.4286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760591
CAS Number: 2807470-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR