6-Bromo-2-chloro-3-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61829-500MG 61829-1G 61829-5G 61829-25G
77331-414EA 233.81 USD
77331-414 77355-662 77367-576 77389-040
6-Bromo-2-chloro-3-propoxybenzaldehyde ≥97%
6-Bromo-2-chloro-3-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClO₂
MW: 277.5422 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760589
CAS Number: 2807443-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR