(3-Chloro-2,4-dimethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

61815-500MG 61815-1G 61815-25G 61815-5G
77331-408EA 304.88 USD
77331-408 77355-658 77389-036 77379-720
(3-Chloro-2,4-dimethoxyphenyl)methanol ≥97%
(3-Chloro-2,4-dimethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34760576
CAS Number: 1593828-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR