3-Chloro-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61813-500MG 61813-1G 61813-5G
77340-134EA 787.95 USD
77340-134 77355-656 77379-718
3-Chloro-2,4-dimethoxybenzoic acid ≥97%
3-Chloro-2,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₄
MW: 216.6183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24494244
CAS Number: 914933-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR