1-Bromo-3-chloro-2-isopropoxy-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61810-500MG 61810-1G 61810-5G 61810-25G
77331-406EA 147.29 USD
77331-406 77345-234 77367-570 77389-034
1-Bromo-3-chloro-2-isopropoxy-4-methoxybenzene ≥97%
1-Bromo-3-chloro-2-isopropoxy-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO₂
MW: 279.5580 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760572
CAS Number: 2764733-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR