1-Bromo-3-chloro-2-ethoxy-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61808-500MG 61808-1G 61808-5G 61808-25G
77331-402EA 145.23 USD
77331-402 77345-104 77367-566 77389-030
1-Bromo-3-chloro-2-ethoxy-4-methoxybenzene ≥97%
1-Bromo-3-chloro-2-ethoxy-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO₂
MW: 265.5315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760570
CAS Number: 2384494-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR