1-Bromo-3-chloro-2,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61807-500MG 61807-1G 61807-5G 61807-25G
77331-400EA 162.74 USD
77331-400 77346-078 77367-564 77389-028
1-Bromo-3-chloro-2,4-dimethoxybenzene ≥97%
1-Bromo-3-chloro-2,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO₂
MW: 251.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618959
CAS Number: 1228956-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR