4-Fluoro-2-((4-methoxybenzyl)oxy)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61804-500MG 61804-1G 61804-5G 61804-25G
77331-398EA 142.14 USD
77331-398 77344-978 77367-562 77389-026
4-Fluoro-2-((4-methoxybenzyl)oxy)-1-methylbenzene ≥97%
4-Fluoro-2-((4-methoxybenzyl)oxy)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₂
MW: 246.2768 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760569
CAS Number: 1979821-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR