1-Chloro-3-((4-methoxybenzyl)oxy)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61802-1G 61802-5G 61802-25G
77340-784EA 60.77 USD
77340-784 77367-560 77379-716
1-Chloro-3-((4-methoxybenzyl)oxy)-2-methylbenzene ≥97%
1-Chloro-3-((4-methoxybenzyl)oxy)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClO₂
MW: 262.7314 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760567
CAS Number: 2764733-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR