4-Bromo-1-((4-methoxybenzyl)oxy)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61801-1G 61801-5G 61801-25G
77340-782EA 60.77 USD
77340-782 77367-558 77379-714
4-Bromo-1-((4-methoxybenzyl)oxy)-2-methylbenzene ≥97%
4-Bromo-1-((4-methoxybenzyl)oxy)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₂
MW: 307.1824 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19496737
CAS Number: 1282915-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR