2-Bromo-4-chloro-1-((4-methoxybenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

61798-1G 61798-5G 61798-25G
77341-810EA 104.03 USD
77341-810 77367-556 77379-712
2-Bromo-4-chloro-1-((4-methoxybenzyl)oxy)benzene ≥97%
2-Bromo-4-chloro-1-((4-methoxybenzyl)oxy)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClO₂
MW: 327.6008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31802731
CAS Number: 2566432-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR