(3-Bromo-4-((4-methoxybenzyl)oxy)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

61795-500MG 61795-1G 61795-5G 61795-25G
77331-396EA 116.39 USD
77331-396 77343-494 77367-554 77379-710
(3-Bromo-4-((4-methoxybenzyl)oxy)phenyl)methanol ≥95%
(3-Bromo-4-((4-methoxybenzyl)oxy)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₃
MW: 323.1818 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19449412
CAS Number: 1280721-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR