(2-Methoxy-5-(trifluoromethyl)phenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

61789-500MG 61789-1G 61789-5G 61789-25G
77331-392EA 152.44 USD
77331-392 77345-474 77367-548 77389-022
(2-Methoxy-5-(trifluoromethyl)phenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(2-Methoxy-5-(trifluoromethyl)phenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₇F₃N₂O₂
MW: 302.2921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760559
CAS Number: 2764728-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR