N,2-dimethoxy-N-methyl-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61788-500MG 61788-1G 61788-5G 61788-25G
77331-390EA 128.75 USD
77331-390 77344-274 77367-546 77379-706
N,2-dimethoxy-N-methyl-5-(trifluoromethyl)benzamide ≥97%
N,2-dimethoxy-N-methyl-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD34760558
CAS Number: 2764733-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR