N,2-dimethoxy-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61787-500MG 61787-1G 61787-5G 61787-25G
77331-388EA 124.63 USD
77331-388 77344-066 77367-544 77379-704
N,2-dimethoxy-5-(trifluoromethyl)benzamide ≥97%
N,2-dimethoxy-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD34760557
CAS Number: 2764729-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR