(2-Chloro-5-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

61784-1G 61784-5G 61784-25G
77341-510EA 90.64 USD
77341-510 77367-540 77379-702
(2-Chloro-5-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(2-Chloro-5-methoxyphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₇ClN₂O₂
MW: 268.7392 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760554
CAS Number: 2807448-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR