(2-Chloro-5-methoxyphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

61783-1G 61783-5G 61783-25G
77341-508EA 90.64 USD
77341-508 77367-538 77379-700
(2-Chloro-5-methoxyphenyl)(thiomorpholino)methanone ≥97%
(2-Chloro-5-methoxyphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClNO₂S
MW: 271.7630 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760553
CAS Number: 2807469-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR