(2-Chloro-5-methoxyphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

61782-1G 61782-5G 61782-25G
77341-506EA 90.64 USD
77341-506 77367-536 77379-698
(2-Chloro-5-methoxyphenyl)(morpholino)methanone ≥97%
(2-Chloro-5-methoxyphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClNO₃
MW: 255.6974 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760552
CAS Number: 2807469-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR