(2-Chloro-5-methoxyphenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

61779-1G 61779-5G 61779-25G
77341-948EA 108.15 USD
77341-948 77367-532 77379-694
(2-Chloro-5-methoxyphenyl)(pyrrolidin-1-yl)methanone ≥97%
(2-Chloro-5-methoxyphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClNO₂
MW: 239.6980 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760549
CAS Number: 2807465-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR