2-Chloro-N-cyclohexyl-5-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

61778-1G 61778-5G 61778-25G
77341-504EA 90.64 USD
77341-504 77367-530 77379-692
2-Chloro-N-cyclohexyl-5-methoxybenzamide ≥97%
2-Chloro-N-cyclohexyl-5-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD34760548
CAS Number: 2807448-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR