2-Chloro-N-cyclopropyl-5-methoxybenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
61777-1G
61777-5G
61777-25G
77342-128EA
114.33
USD
77342-128
77367-528
77379-690
2-Chloro-N-cyclopropyl-5-methoxybenzamide ≥97%
2-Chloro-N-cyclopropyl-5-methoxybenzamide
Precaución: for Research Use Only
Formula:
C₁₁H₁₂ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD34760547 CAS Number: 2807470-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...