2-Chloro-5-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

61776-1G 61776-5G 61776-25G
77342-078EA 112.27 USD
77342-078 77367-526 77379-688
2-Chloro-5-methoxy-N-methylbenzamide ≥97%
2-Chloro-5-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD18905097
CAS Number: 1203954-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR