2-Chloro-5-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

61775-500MG 61775-1G 61775-5G 61775-25G
77331-382EA 92.7 USD
77331-382 77342-468 77367-524 77379-686
2-Chloro-5-methoxybenzamide ≥97%
2-Chloro-5-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393557
CAS Number: 62798-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR