1-Chloro-3-ethoxy-2-fluoro-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

61738-500MG 61738-1G 61738-5G 61738-25G
77331-376EA 136.99 USD
77331-376 77344-552 77367-516 77389-016
1-Chloro-3-ethoxy-2-fluoro-5-(trifluoromethyl)benzene ≥95%
1-Chloro-3-ethoxy-2-fluoro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD34760524
CAS Number: 2386070-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR