(2,4-Dichloro-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

61736-500MG 61736-1G 61736-5G
77338-190EA 468.65 USD
77338-190 77355-650 77379-680
(2,4-Dichloro-5-methylphenyl)(methyl)sulfane ≥97%
(2,4-Dichloro-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂S
MW: 207.1201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787039
CAS Number: 1806275-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR