1-(2,4-Dichloro-5-methylphenyl)-2,2,2-trifluoroethanol ≥97%

Supplier: AOBChem USA

Warning

61735-500MG 61735-1G 61735-5G
77336-436EA 387.28 USD
77336-436 77355-648 77379-678
1-(2,4-Dichloro-5-methylphenyl)-2,2,2-trifluoroethanol ≥97%
1-(2,4-Dichloro-5-methylphenyl)-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₉H₇Cl₂F₃O
MW: 259.0524 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760522
CAS Number: 2764733-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR