1-(2,4-Dichloro-5-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

61734-500MG 61734-1G 61734-5G 61734-25G
77331-374EA 223.51 USD
77331-374 77355-646 77367-514 77389-014
1-(2,4-Dichloro-5-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,4-Dichloro-5-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅Cl₂F₃O
MW: 257.0366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760521
CAS Number: 2764731-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR