2-(2,4-Difluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

61731-500MG 61731-1G 61731-5G 61731-25G
77331-372EA 250.29 USD
77331-372 77355-642 77367-512 77389-012
2-(2,4-Difluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2,4-Difluoro-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₂
MW: 254.0806 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19440199
CAS Number: 1427407-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR