5-(tert-Butyl)-2-chloro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61724-500MG 61724-1G
77340-576EA 2424.62 USD
77340-576 77355-638
5-(tert-Butyl)-2-chloro-3-formylbenzoic acid ≥97%
5-(tert-Butyl)-2-chloro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClO₃
MW: 240.6828 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760513
CAS Number: 2807446-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR