1-Bromo-5-(tert-butyl)-2-chloro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

61718-500MG 61718-1G 61718-5G
77339-376EA 582.98 USD
77339-376 77355-634 77379-670
1-Bromo-5-(tert-butyl)-2-chloro-3-iodobenzene ≥97%
1-Bromo-5-(tert-butyl)-2-chloro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrClI
MW: 373.4558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760507
CAS Number: 2764731-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR