3-Bromo-5-(tert-butyl)-2-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61713-500MG 61713-1G 61713-5G
77336-902EA 398.61 USD
77336-902 77355-630 77379-666
3-Bromo-5-(tert-butyl)-2-chlorobenzoic acid ≥97%
3-Bromo-5-(tert-butyl)-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrClO₂
MW: 291.5687 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760502
CAS Number: 2918939-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR