3',4'-Difluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

61702-500MG 61702-1G 61702-5G 61702-25G
77331-368EA 125.66 USD
77331-368 77344-096 77367-506 77379-660
3',4'-Difluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%
3',4'-Difluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈F₂O
MW: 218.1988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06858701
CAS Number: 343604-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR