3,4-Difluoro-4'-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61701-500MG 61701-1G 61701-5G
77331-366EA 318.27 USD
77331-366 77355-626 77379-658
3,4-Difluoro-4'-methoxy-1,1'-biphenyl ≥97%
3,4-Difluoro-4'-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂O
MW: 220.2147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760493
CAS Number: 182925-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR