3',4'-Difluoro-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61700-500MG 61700-1G 61700-5G 61700-25G
77331-364EA 183.34 USD
77331-364 77347-128 77367-504 77389-008
3',4'-Difluoro-2-methyl-1,1'-biphenyl ≥97%
3',4'-Difluoro-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂
MW: 204.2153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760492
CAS Number: 2764731-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR