3,4-Difluoro-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61699-500MG 61699-1G 61699-5G 61699-25G
77331-362EA 208.06 USD
77331-362 77355-624 77367-502 77389-006
3,4-Difluoro-4'-methyl-1,1'-biphenyl ≥97%
3,4-Difluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂
MW: 204.2153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760491
CAS Number: 182169-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR