3-(3,4-Difluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

61698-500MG 61698-1G 61698-5G 61698-25G
77331-360EA 212.18 USD
77331-360 77355-622 77367-500 77389-004
3-(3,4-Difluorophenyl)pyridine ≥97%
3-(3,4-Difluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇F₂N
Storage Temperature: Ambient
MDL Number: MFCD15476737
CAS Number: 1214376-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR