2,4,4'-Trifluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

61694-500MG 61694-1G 61694-5G 61694-25G
77331-356EA 244.11 USD
77331-356 77355-620 77367-496 77389-000
2,4,4'-Trifluoro-1,1'-biphenyl ≥95%
2,4,4'-Trifluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇F₃
MW: 208.1791 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760490
CAS Number: 872545-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR