2',4'-Difluoro-2,3-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

61692-500MG 61692-1G 61692-5G 61692-25G
77331-354EA 163.77 USD
77331-354 77346-120 77367-494 77388-998
2',4'-Difluoro-2,3-dimethyl-1,1'-biphenyl ≥95%
2',4'-Difluoro-2,3-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂
MW: 218.2418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760489
CAS Number: 2764729-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR