2,4-Difluoro-2'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61691-500MG 61691-1G 61691-5G 61691-25G
77331-352EA 157.59 USD
77331-352 77345-652 77367-492 77388-996
2,4-Difluoro-2'-methyl-1,1'-biphenyl ≥97%
2,4-Difluoro-2'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂
Storage Temperature: Ambient
MDL Number: MFCD34760488
CAS Number: 1432742-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR