2,4-Difluoro-4'-methoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

61690-500MG 61690-1G 61690-5G 61690-25G
77331-350EA 182.31 USD
77331-350 77346-980 77367-490 77388-994
2,4-Difluoro-4'-methoxy-1,1'-biphenyl ≥95%
2,4-Difluoro-4'-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂O
MW: 220.2147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23099747
CAS Number: 90101-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR