4-(2,4-Difluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

61688-500MG 61688-1G 61688-5G 61688-25G
77331-346EA 208.06 USD
77331-346 77355-616 77367-486 77388-990
4-(2,4-Difluorophenyl)pyridine ≥97%
4-(2,4-Difluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇F₂N
MW: 191.1767 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864842
CAS Number: 914349-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR