3-Bromo-4'-methoxy-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
61686-500MG
61686-1G
61686-5G
61686-25G
77331-342EA
198.79
USD
77331-342
77355-612
77367-482
77388-986
3-Bromo-4'-methoxy-1,1'-biphenyl ≥97%
3-Bromo-4'-methoxy-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₃H₁₁BrO MW: 263.1298 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06201390 CAS Number: 74447-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...