3'-Bromo-2,3-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61685-500MG 61685-1G 61685-5G 61685-25G
77331-340EA 226.6 USD
77331-340 77355-610 77367-480 77388-984
3'-Bromo-2,3-dimethyl-1,1'-biphenyl ≥97%
3'-Bromo-2,3-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br
MW: 261.1570 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14582784
CAS Number: 1443309-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR