4'-Bromo-2'-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

61682-500MG 61682-1G 61682-5G 61682-25G
77331-336EA 183.34 USD
77331-336 77347-126 77367-478 77388-980
4'-Bromo-2'-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%
4'-Bromo-2'-fluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈BrFO
MW: 279.1044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34641985
CAS Number: 2764729-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR