3-Chloro-4-ethoxy-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61678-500MG 61678-1G 61678-5G
77338-986EA 540.75 USD
77338-986 77355-604 77379-652
3-Chloro-4-ethoxy-2-methoxybenzaldehyde ≥97%
3-Chloro-4-ethoxy-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.6455 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760484
CAS Number: 2386253-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR